Product Details
- SNP ID
-
rs734358
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:41480241 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTATATGTTGGGTCCCGTCCATCCC[A/C]CCATCCTGCCTTTTCTCCTGTTTTT
- Phenotype
-
MIM: 131241
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EDN2
PubMed Links
Gene Details
- Gene
- EDN2
- Gene Name
- endothelin 2
View Full Product Details