Product Details

SNP ID
rs708119
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:228015567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGCAGGCTGGCCTCAGCCTCT[C/G]CCCGGGGACTGGGCCCAGCCTTCCT
Phenotype
MIM: 606359
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WNT3A PubMed Links

Gene Details

Gene
WNT3A
Gene Name
Wnt family member 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033131.3 Intron NP_149122.1
XM_011544319.2 Intron XP_011542621.1

View Full Product Details