Product Details

SNP ID
rs642845
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:62442564 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGTATACTATAATCACAATATGT[C/T]ATACCCATGTACAACTTATTTACCC
Phenotype
MIM: 603478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP1 PubMed Links

Gene Details

Gene
USP1
Gene Name
ubiquitin specific peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017415.1 Intron NP_001017415.1
NM_001017416.1 Intron NP_001017416.1
NM_003368.4 Intron NP_003359.3

View Full Product Details