Product Details
- SNP ID
-
rs534030
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:197767074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACATGTCTATTTTTTAATTTACC[A/C]CCTTTTTTTTTTTAACGGGCAGCCC
- Phenotype
-
MIM: 616933
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FYTTD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs536180798] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FYTTD1
- Gene Name
- forty-two-three domain containing 1
View Full Product Details