Product Details

SNP ID
rs717025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50350542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCACTTAATACTCATTTCTCTT[C/T]ATCCAGGAAGGCAAAAACTATTATC
Phenotype
MIM: 600610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ10038 PubMed Links

Gene Details

Gene
FLJ10038
Gene Name
uncharacterized protein FLJ10038
There are no transcripts associated with this gene.

Gene
GABPB1
Gene Name
GA binding protein transcription factor beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320910.1 Intron NP_001307839.1
NM_001320915.1 Intron NP_001307844.1
NM_002041.4 Intron NP_002032.2
NM_005254.5 Intron NP_005245.2
NM_016654.4 Intron NP_057738.1
NM_016655.4 Intron NP_057739.1
NM_181427.3 Intron NP_852092.1
XM_005254274.3 Intron XP_005254331.1
XM_011521426.2 Intron XP_011519728.1
XM_017022053.1 Intron XP_016877542.1
XM_017022054.1 Intron XP_016877543.1
Gene
GABPB1-AS1
Gene Name
GABPB1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MIR4712
Gene Name
microRNA 4712
There are no transcripts associated with this gene.

View Full Product Details