Product Details

SNP ID
rs537386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66684287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCCTGAAAGCTCCGATTGCC[A/C]AACCAGAGACAAAGGAGAAGCCACC
Phenotype
MIM: 604985
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RBM4B PubMed Links

Gene Details

Gene
RBM4B
Gene Name
RNA binding motif protein 4B
There are no transcripts associated with this gene.

Gene
SPTBN2
Gene Name
spectrin beta, non-erythrocytic 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006946.2 9583 Intron NP_008877.1
XM_005274192.4 9583 UTR 3 XP_005274249.1
XM_005274193.3 9583 UTR 3 XP_005274250.1
XM_006718669.3 9583 UTR 3 XP_006718732.1
XM_006718671.3 9583 UTR 3 XP_006718734.1
XM_011545216.2 9583 Intron XP_011543518.1
XM_011545217.2 9583 Intron XP_011543519.1
XM_017018174.1 9583 UTR 3 XP_016873663.1
XM_017018175.1 9583 UTR 3 XP_016873664.1
XM_017018176.1 9583 UTR 3 XP_016873665.1
XM_017018177.1 9583 UTR 3 XP_016873666.1
XM_017018178.1 9583 UTR 3 XP_016873667.1

View Full Product Details