Product Details

SNP ID
rs201897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32606104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATTGCAGTAGGCTAACAATCT[A/T]ATGCTGAAAACAAGAATGAAAAACA
Phenotype
MIM: 612328 MIM: 609641
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC73 PubMed Links

Gene Details

Gene
CCDC73
Gene Name
coiled-coil domain containing 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008391.3 Intron NP_001008392.2
XM_011520139.2 Intron XP_011518441.1
Gene
EIF3M
Gene Name
eukaryotic translation initiation factor 3 subunit M
There are no transcripts associated with this gene.

View Full Product Details