Product Details

SNP ID
rs325917
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:111118906 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGAAGCACATAACCCTGAGTGA[C/T]GGGAAAAAAAGAATAGTTTTGTGAA
Phenotype
MIM: 613360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRAM2 PubMed Links

Gene Details

Gene
DRAM2
Gene Name
DNA damage regulated autophagy modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178454.4 Intron NP_848549.3
XM_005270469.2 Intron XP_005270526.1
XM_005270470.1 Intron XP_005270527.1
XM_006710361.1 Intron XP_006710424.1
XM_006710362.1 Intron XP_006710425.1
XM_011540707.1 Intron XP_011539009.1
XM_011540708.1 Intron XP_011539010.1
XM_017000320.1 Intron XP_016855809.1
XM_017000321.1 Intron XP_016855810.1

View Full Product Details