Product Details

SNP ID
rs353312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65565983 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTTTAACATGTTCTTTGGACAT[G/T]GGACTGTTGTATATATTTTCAGAAA
Phenotype
MIM: 611502
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CENPK PubMed Links

Gene Details

Gene
CENPK
Gene Name
centromere protein K
There are no transcripts associated with this gene.

Gene
PPWD1
Gene Name
peptidylprolyl isomerase domain and WD repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278926.1 Intron NP_001265855.1
NM_001278927.1 Intron NP_001265856.1
NM_001278929.1 Intron NP_001265858.1
NM_015342.3 Intron NP_056157.1
XM_005248474.3 Intron XP_005248531.1
XM_006714583.3 Intron XP_006714646.1
XM_011543295.1 Intron XP_011541597.1
XM_017009286.1 Intron XP_016864775.1

View Full Product Details