Product Details

SNP ID
rs134757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26170791 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCTTGCAAGGTCCCAGAAATGC[G/T]CTCCTCACGTGTAAAAAAGCAGACA
Phenotype
MIM: 607021
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEZ6L PubMed Links

Gene Details

Gene
SEZ6L
Gene Name
seizure related 6 homolog like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184773.1 Intron NP_001171702.1
NM_001184774.1 Intron NP_001171703.1
NM_001184775.1 Intron NP_001171704.1
NM_001184776.1 Intron NP_001171705.1
NM_001184777.1 Intron NP_001171706.1
NM_021115.4 Intron NP_066938.2
XM_005261439.3 Intron XP_005261496.1
XM_005261440.3 Intron XP_005261497.1
XM_006724195.3 Intron XP_006724258.1
XM_011530037.2 Intron XP_011528339.1
XM_011530038.2 Intron XP_011528340.1
XM_011530039.2 Intron XP_011528341.1
XM_017028699.1 Intron XP_016884188.1
XM_017028700.1 Intron XP_016884189.1

View Full Product Details