Product Details
- SNP ID
-
rs175444
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:75135079 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTGGCTAAACAATGGCAGGGGAG[C/G]TGGGTAAAGGCAATTAATTAACTTC
- Phenotype
-
MIM: 609798
MIM: 605406
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NEK9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141521366] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NEK9
- Gene Name
- NIMA related kinase 9
There are no transcripts associated with this gene.
- Gene
- TMED10
- Gene Name
- transmembrane p24 trafficking protein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006827.5 |
|
Intron |
|
|
NP_006818.3 |
View Full Product Details