Product Details

SNP ID
rs4077439
Assay Type
Functionally tested
NCBI dbSNP Submissions
70
Location
Chr.1:33269760 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGGCGGGGGTTGGGGGATGGCC[A/G]TAAGTTGCTTTACTGAACCCTTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF362 PubMed Links

Gene Details

Gene
ZNF362
Gene Name
zinc finger protein 362
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152493.2 36 Intron NP_689706.2
XM_005270507.2 36 Intron XP_005270564.1
XM_005270508.4 36 UTR 5 XP_005270565.1
XM_017000415.1 36 UTR 5 XP_016855904.1

View Full Product Details