Product Details

SNP ID
rs2465413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62629625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGCAGGTTCCTTCCCTCGGAAC[C/T]GTGCCCTCTGCACAGAGTGCGCTTG
Phenotype
MIM: 612264
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRC2 PubMed Links

Gene Details

Gene
MRC2
Gene Name
mannose receptor C type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006039.4 Intron NP_006030.2
XM_011525543.2 Intron XP_011523845.1

View Full Product Details