Product Details

SNP ID
rs6758667
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47088817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCTGGAAATCTAATCAAGTCCT[A/G]GCCAAACGTCAAAGGAAGCTGAAGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C2orf61 PubMed Links
Additional Information
For this assay, SNP(s) [rs75828512] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C2orf61
Gene Name
chromosome 2 open reading frame 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163561.1 Intron NP_001157033.1
NM_173649.2 Intron NP_775920.1

View Full Product Details