Product Details

SNP ID
rs1524026
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:227877929 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTTTGTCACAGCGAAGCAGTCAAA[C/T]GTACATGTTTCAATTTTGTAGTGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAW1 PubMed Links

Gene Details

Gene
DAW1
Gene Name
dynein assembly factor with WD repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011510755.1 Intron XP_011509057.1
XM_017003490.1 Intron XP_016858979.1

View Full Product Details