Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spe-8
Entrez Gene ID
171606
Gene Name
Spermatocyte protein spe-8
Gene Aliases
CELE_F53G12.6
Chromosome Location
Chr.I: 118093 - 120901 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058279.2 NP_490680.1 1 - 2 117 60

View Full Product Details