Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.


View Full Product Details