Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egrh-1
Entrez Gene ID
181580
Gene Name
EGR (Early Growth factor Response factor) Homolog
Gene Aliases
CELE_C27C12.2
Chromosome Location
Chr.X: 14845937 - 14849217 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078061.6 NP_510462.2 2 - 3 510 110

View Full Product Details