Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-6
Entrez Gene ID
181584
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_F31F6.5
Chromosome Location
Chr.X: 14888488 - 14894333 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078071.7 NP_510472.2 7 - 8 1265 69

View Full Product Details