Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mua-6
Entrez Gene ID
181698
Gene Name
Intermediate filament protein ifa-2
Gene Aliases
CELE_W10G6.3
Chromosome Location
Chr.X: 16262289 - 16265686 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078247.9 NP_510648.1 1 - 2 476 75

View Full Product Details