Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-10
Entrez Gene ID
24104866
Gene Name
Probable medium-chain specific acyl-CoA dehydrogenase 10, mitochondrial
Gene Aliases
CELE_T08G2.3
Chromosome Location
Chr.X: 17213144 - 17214903 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307895.2 NP_001294824.1 4 - 5 1095 96

View Full Product Details