Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crb-1
Entrez Gene ID
181771
Gene Name
Drosophila CRumBs homolog
Gene Aliases
CELE_F11C7.4
Chromosome Location
Chr.X: 17402532 - 17414014 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078421.6 NP_510822.1 19 - 20 3305 89

View Full Product Details