Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cdc-26
- Entrez Gene ID
-
172958
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_B0511.9
- Chromosome Location
-
Chr.I: 10639076 - 10640234 on Build WBcel235
- Gene Symbol
- mrps-30
- Entrez Gene ID
-
172957
- Gene Name
- Mitochondrial Ribosomal Protein, Small
- Gene Aliases
-
CELE_B0511.8
- Chromosome Location
-
Chr.I: 10636687 - 10640153 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060382.7 |
NP_492783.1 |
1 - 2 |
127 |
71 |
View Full Product Details