Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-38
Entrez Gene ID
174136
Gene Name
protein-coding
Gene Aliases
CELE_F22D3.1
Chromosome Location
Chr.II: 6951526 - 6955409 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351905.4 NP_001338814.1 4 - 4 705 97
NM_001351913.2 NP_001338820.1 4 - 5 587 97
NM_001351914.2 NP_001338821.1 4 - 5 696 97
NM_001381471.2 NP_001368648.1 4 - 5 570 97
NM_001381472.2 NP_001368657.1 4 - 5 604 97
NM_001381473.2 NP_001368649.1 3 - 4 526 97
NM_001381474.1 NP_001368650.1 3 - 4 460 97
NM_001381475.2 NP_001368651.1 4 - 5 612 97
NM_001381476.1 NP_001368652.1 3 - 4 532 97
NM_001381477.2 NP_001367332.1 4 - 5 556 97
NM_001381478.2 NP_001368653.1 3 - 4 523 97
NM_001381479.1 NP_001368654.1 3 - 4 518 97
NM_001381480.1 NP_001368655.1 3 - 4 523 97
NM_001381481.1 NP_001368656.1 3 - 4 451 97
NM_001383875.1 4 - 5 705 97
NM_171852.2 NP_741018.1 4 - 5 582 97

View Full Product Details