Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-19
Entrez Gene ID
177513
Gene Name
protein-coding
Gene Aliases
CELE_C48A7.1
Chromosome Location
Chr.IV: 7405582 - 7418740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027908.6 NP_001023079.1 10 - 11 4491 122
NM_171379.6 NP_741442.1 10 - 11 4491 122
GenBank mRNA AF023602.1 - 11 - 12 4524 122

View Full Product Details