Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsp-6
Entrez Gene ID
177566
Gene Name
Probable splicing factor, arginine/serine-rich 6
Gene Aliases
CELE_C33H5.12
Chromosome Location
Chr.IV: 7789845 - 7791229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171383.5 NP_741446.1 4 - 5 449 82
NR_131573.1 5 - 6 695 82
NR_131574.1 3 - 4 887 82

View Full Product Details