Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chn-1
Entrez Gene ID
172303
Gene Name
C-term of Hsp70-iNteracting protein (CHIP family)
Gene Aliases
CELE_T09B4.10
Chromosome Location
Chr.I: 6181314 - 6185197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059380.8 NP_491781.2 7 - 8 801 93
GenBank mRNA AF303269.1 - 6 - 7 746 93

View Full Product Details