Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-56
Entrez Gene ID
188271
Gene Name
COLlagen
Gene Aliases
CELE_T08B2.2
Chromosome Location
Chr.I: 6232777 - 6234280 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059399.1 NP_491800.1 3 - 4 700 66

View Full Product Details