Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-19
Entrez Gene ID
192069
Gene Name
TetraSPanin family
Gene Aliases
CELE_D2092.7
Chromosome Location
Chr.I: 6616304 - 6618560 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374906.1 NP_001361987.1 5 - 6 771 65

View Full Product Details