Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ser-3
Entrez Gene ID
259744
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K02F2.6
Chromosome Location
Chr.I: 6820517 - 6823093 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059553.5 NP_491954.1 4 - 5 1237 67

View Full Product Details