Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-3
Entrez Gene ID
172449
Gene Name
GLutamate Receptor family (AMPA)
Gene Aliases
CELE_K10D3.1
Chromosome Location
Chr.I: 7117808 - 7121545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059616.3 NP_492017.3 12 - 13 1966 78

View Full Product Details