Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-13
Entrez Gene ID
172497
Gene Name
protein-coding
Gene Aliases
CELE_ZK524.2
Chromosome Location
Chr.I: 7422785 - 7454116 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026700.2 NP_001021871.1 1 - 2 63 66
NM_001026702.3 NP_001021873.1 1 - 2 63 66
NM_001026703.4 NP_001021874.1 1 - 2 63 66
NM_001263573.3 NP_001250502.1 1 - 2 63 66
NM_001330927.3 NP_001317864.1 1 - 2 63 66

View Full Product Details