Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- F26A3.1
- Entrez Gene ID
-
172522
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F26A3.1
- Chromosome Location
-
Chr.I: 7640340 - 7643330 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059728.8 |
NP_492129.2 |
2 - 3 |
337 |
89 |
- Gene Symbol
- ncbp-2
- Entrez Gene ID
-
184953
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F26A3.2
- Chromosome Location
-
Chr.I: 7641883 - 7643328 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383376.1 |
|
2 - 3 |
335 |
89 |
View Full Product Details