Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
del-4
Entrez Gene ID
189033
Gene Name
DEgenerin Like
Gene Aliases
CELE_T28B8.5
Chromosome Location
Chr.I: 8139465 - 8142537 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059829.6 NP_492230.2 7 - 8 1005 104

View Full Product Details