Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-36A1
Entrez Gene ID
183193
Gene Name
Probable cytochrome P450 CYP36A1
Gene Aliases
CELE_C34B7.3
Chromosome Location
Chr.I: 8341681 - 8346023 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059866.6 NP_492267.1 7 - 8 1256 84

View Full Product Details