Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sig-7
Entrez Gene ID
172664
Gene Name
protein-coding
Gene Aliases
CELE_F39H2.2
Chromosome Location
Chr.I: 8646451 - 8651781 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373191.2 NP_001359651.1 6 - 7 851 145
NM_059942.8 NP_492343.2 6 - 7 987 145

View Full Product Details