Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syp-3
Entrez Gene ID
172667
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F39H2.4
Chromosome Location
Chr.I: 8663045 - 8664445 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059944.8 NP_492345.2 4 - 5 476 140

View Full Product Details