Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-7
Entrez Gene ID
186153
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_F53B6.6
Chromosome Location
Chr.I: 8954559 - 8957394 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374976.1 NP_001361852.1 2 - 3 153 79

View Full Product Details