Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-19
Entrez Gene ID
191605
Gene Name
protein-coding
Gene Aliases
CELE_C31H5.3
Chromosome Location
Chr.I: 9031235 - 9036695 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136284.4 NP_001129756.1 5 - 6 903 72
NM_060028.7 NP_492429.2 5 - 6 904 72
GenBank mRNA AY523514.1 - 5 - 6 904 72

View Full Product Details