Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prom-1
Entrez Gene ID
185001
Gene Name
protein-coding
Gene Aliases
CELE_F26H9.1
Chromosome Location
Chr.I: 9287443 - 9289770 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060076.8 NP_492477.2 2 - 3 247 120

View Full Product Details