Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cri-1
Entrez Gene ID
187082
Gene Name
Headcase domain-containing protein
Gene Aliases
CELE_K07A1.7
Chromosome Location
Chr.I: 9594181 - 9598363 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060146.3 NP_492547.2 3 - 4 603 69

View Full Product Details