Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-49
Entrez Gene ID
172839
Gene Name
protein-coding
Gene Aliases
CELE_K10C3.6
Chromosome Location
Chr.I: 9869632 - 9874680 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381165.1 NP_001367039.1 3 - 4 311 84
NM_001381168.2 NP_001366915.1 3 - 4 397 84
NM_060212.7 NP_492613.1 3 - 4 378 84
NM_181999.6 NP_871799.1 3 - 4 266 84

View Full Product Details