Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oct-1
Entrez Gene ID
172841
Gene Name
protein-coding
Gene Aliases
CELE_F52F12.1
Chromosome Location
Chr.I: 9888356 - 9892223 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026311.7 NP_001021482.1 2 - 3 158 70
NM_060216.6 NP_492617.4 3 - 4 221 70
GenBank mRNA AF110415.1 - 2 - 3 166 70

View Full Product Details