Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sys-1
Entrez Gene ID
172859
Gene Name
protein-coding
Gene Aliases
CELE_T23D8.9
Chromosome Location
Chr.I: 9980063 - 9984794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383706.1 3 - 4 574 143

View Full Product Details