Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pab-1
Entrez Gene ID
172920
Gene Name
Polyadenylate-binding protein
Gene Aliases
CELE_Y106G6H.2
Chromosome Location
Chr.I: 10432815 - 10435778 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383718.1 2 - 3 537 71

View Full Product Details