Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sec-8
- Entrez Gene ID
-
172924
- Gene Name
- Exocyst complex component 4
- Gene Aliases
-
CELE_Y106G6H.7
- Chromosome Location
-
Chr.I: 10446657 - 10471767 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060331.8 |
NP_492732.2 |
11 - 12 |
1500 |
135 |
- Gene Symbol
- Y106G6H.8
- Entrez Gene ID
-
190926
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y106G6H.8
- Chromosome Location
-
Chr.I: 10449459 - 10452896 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383720.1 |
|
1 - 2 |
94 |
135 |
View Full Product Details