Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cutl-13
Entrez Gene ID
171637
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_W04C9.3
Chromosome Location
Chr.I: 477582 - 489242 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058335.5 NP_490736.2 4 - 5 773 90

View Full Product Details