Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pif-1
Entrez Gene ID
171665
Gene Name
protein-coding
Gene Aliases
CELE_Y18H1A.6
Chromosome Location
Chr.I: 719929 - 731078 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306244.3 NP_001293173.1 4 - 5 468 140
NM_001306245.3 NP_001293174.1 8 - 9 999 140
NM_058373.6 NP_490774.2 10 - 11 1466 140
GenBank mRNA AB015041.1 - 10 - 11 1323 140

View Full Product Details