Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpa-16
Entrez Gene ID
171675
Gene Name
protein-coding
Gene Aliases
CELE_Y95B8A.5
Chromosome Location
Chr.I: 897286 - 907370 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058389.4 NP_490790.1 1 - 2 70 72
GenBank mRNA AY008138.1 - 1 - 2 66 72

View Full Product Details