Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spd-1
Entrez Gene ID
171683
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y34D9A.4
Chromosome Location
Chr.I: 1030632 - 1034631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058408.6 NP_490809.2 2 - 3 246 80
GenBank mRNA AY714241.1 - 1 - 2 185 80

View Full Product Details